Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078934.3 | NP_523658.1 | 2 - 3 | 514 | 66 | |
GenBank mRNA | AF178978.1 | - | 2 - 3 | 499 | 66 | |
AY069670.1 | - | 2 - 3 | 495 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |