Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078934.3 | NP_523658.1 | 3 - 4 | 672 | 103 | |
GenBank mRNA | AF178978.1 | - | 3 - 4 | 657 | 103 | |
AY069670.1 | - | 3 - 4 | 653 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |