Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078934.3 | NP_523658.1 | 6 - 7 | 1587 | 80 | |
GenBank mRNA | AF178978.1 | - | 6 - 7 | 1572 | 80 | |
AY069670.1 | - | 6 - 7 | 1568 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |