Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273926.1 | NP_001260855.1 | 3 - 4 | 868 | 76 | |
NM_001273927.2 | NP_001260856.1 | 3 - 4 | 868 | 76 | ||
NM_001299345.1 | NP_001286274.1 | 3 - 4 | 1301 | 76 | ||
NM_078955.3 | NP_523679.2 | 2 - 3 | 838 | 76 | ||
GenBank mRNA | BT133422.1 | - | 2 - 3 | 826 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |