Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001370044.1 | NP_001356961.1 | 3 - 4 | 963 | 106 | |
NM_078963.3 | NP_523687.1 | 3 - 4 | 963 | 106 | ||
GenBank mRNA | AB112932.1 | - | 2 - 3 | 344 | 106 | |
AY060449.1 | - | 3 - 4 | 950 | 106 | ||
M64621.1 | - | 3 - 4 | 930 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |