Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zetaTry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078967.4 | NP_523691.1 | 1 | 895 | 79 | |
GenBank mRNA | AY113523.1 | - | 1 | 891 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |