Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: iotaTry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078971.4 | NP_523695.1 | 1 | 763 | 61 | |
GenBank mRNA | AY113550.1 | - | 1 | 761 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |