Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169613.1 | NP_001163084.1 | 3 - 4 | 957 | 90 | |
NM_136508.4 | NP_610352.2 | 3 - 4 | 961 | 90 | ||
GenBank mRNA | BT021374.1 | - | 3 - 4 | 915 | 90 | |
BT050475.1 | - | 3 - 4 | 908 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |