Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169613.1 | NP_001163084.1 | 9 - 10 | 2607 | 164 | |
NM_136508.4 | NP_610352.2 | 9 - 10 | 2611 | 164 | ||
GenBank mRNA | BT021374.1 | - | 9 - 10 | 2565 | 164 | |
BT050475.1 | - | 9 - 10 | 2558 | 164 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |