Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014505.2 | NP_001014505.1 | 3 - 4 | 1601 | 87 | |
GenBank mRNA | BT050478.1 | - | 3 - 4 | 1601 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |