Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: boly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136529.4 | NP_610373.2 | 1 | 1064 | 90 | |
GenBank mRNA | AY118800.1 | - | 1 | 1036 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |