Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4ad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273876.1 | NP_001260805.1 | 2 - 3 | 275 | 84 | |
NM_136536.3 | NP_610380.1 | 4 - 5 | 680 | 84 | ||
GenBank mRNA | AY061058.1 | - | 4 - 5 | 653 | 84 | |
BT133029.1 | - | 2 - 3 | 275 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |