Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hydr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201963.2 | NP_001188892.1 | 2 - 3 | 589 | 86 | |
NM_136615.4 | NP_610459.2 | 2 - 3 | 411 | 86 | ||
GenBank mRNA | BT150340.1 | - | 2 - 3 | 354 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |