Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4p2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136628.3 | NP_610472.1 | 3 - 4 | 528 | 86 | |
GenBank mRNA | AY051564.1 | - | 3 - 4 | 510 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |