Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273892.1 | NP_001260821.1 | 4 - 5 | 2101 | 112 | |
NM_001299317.1 | NP_001286246.1 | 4 - 5 | 1767 | 112 | ||
NM_136635.3 | NP_610479.1 | 4 - 5 | 2101 | 112 | ||
GenBank mRNA | AF422803.1 | - | 4 - 5 | 1744 | 112 | |
AY069468.1 | - | 4 - 5 | 2046 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |