Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144152.2 | NP_001137624.1 | 15 - 16 | 5418 | 101 | |
NM_001299320.1 | NP_001286249.1 | 13 - 14 | 4026 | 101 | ||
NM_136647.5 | NP_610491.4 | 13 - 14 | 4697 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |