Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir47a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136736.1 | NP_610580.1 | 1 | 2935 | 110 | |
GenBank mRNA | BT031294.1 | - | 1 | 2935 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |