Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136839.3 | NP_610683.1 | 2 - 3 | 648 | 59 | |
GenBank mRNA | AY051563.1 | - | 2 - 3 | 627 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |