Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cuff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136865.3 | NP_610709.1 | 3 - 4 | 935 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |