Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pds5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259342.2 | NP_001246271.1 | 7 - 8 | 2370 | 69 | |
NM_136875.4 | NP_610719.1 | 7 - 8 | 2112 | 69 | ||
GenBank mRNA | BT053744.1 | - | 7 - 8 | 2092 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |