Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273980.2 | NP_001260909.1 | 3 - 4 | 789 | 73 | |
NM_136877.5 | NP_610721.2 | 3 - 4 | 789 | 73 | ||
GenBank mRNA | BT010245.1 | - | 3 - 4 | 786 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |