Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273981.1 | NP_001260910.1 | 1 - 2 | 2742 | 65 | |
NM_136882.3 | NP_610726.1 | 1 - 2 | 2255 | 65 | ||
GenBank mRNA | AF425733.1 | - | 1 - 2 | 2256 | 65 | |
AF425734.1 | - | 2 - 3 | 3596 | 65 | ||
AY060310.1 | - | 1 - 2 | 2274 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |