Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha36-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136909.3 | NP_610753.1 | 1 - 2 | 595 | 84 | |
GenBank mRNA | BT132958.1 | - | 1 - 2 | 595 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |