Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr49Ae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136930.4 | NP_610774.3 | 2 - 3 | 241 | 68 | |
GenBank mRNA | AY061515.1 | - | 2 - 3 | 219 | 68 | |
KX531749.1 | - | 1 - 2 | 135 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |