Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: TBPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169809.2 | NP_001163280.1 | 6 - 7 | 1874 | 69 | |
NM_001169810.2 | NP_001163281.1 | 6 - 7 | 1645 | 69 | ||
NM_058051.4 | NP_477399.1 | 6 - 7 | 1401 | 69 | ||
NM_058052.4 | NP_477400.1 | 6 - 7 | 1642 | 69 | ||
NM_134318.3 | NP_599145.1 | 5 - 6 | 2674 | 69 | ||
NM_166628.3 | NP_726373.1 | 6 - 7 | 1871 | 69 | ||
GenBank mRNA | AY089497.1 | - | 6 - 7 | 1630 | 69 | |
BT022083.1 | - | 6 - 7 | 1894 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |