Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yki | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043103.2 | NP_001036568.2 | 1 - 2 | 1025 | 86 | |
NM_001363931.1 | NP_001350857.1 | 1 - 2 | 1025 | 86 | ||
NM_138035.3 | NP_611879.4 | 1 - 2 | 1025 | 86 | ||
NM_166657.5 | NP_726414.3 | 2 - 3 | 960 | 86 | ||
GenBank mRNA | BT132826.1 | - | 1 - 2 | 946 | 86 | |
DQ099897.1 | - | 1 - 2 | 1025 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |