Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169824.2 | NP_001163295.1 | 3 - 4 | 938 | 67 | |
NM_001299920.1 | NP_001286849.1 | 3 - 4 | 184 | 67 | ||
NM_138094.4 | NP_611938.2 | 4 - 5 | 462 | 67 | ||
NM_166677.3 | NP_726461.1 | 4 - 5 | 466 | 67 | ||
GenBank mRNA | AY862157.1 | - | 4 - 5 | 436 | 67 | |
BT003795.1 | - | 3 - 4 | 921 | 67 | ||
BT011382.1 | - | 4 - 5 | 434 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |