Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pio | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169824.2 | NP_001163295.1 | 7 - 8 | 2036 | 66 | |
NM_001299920.1 | NP_001286849.1 | 7 - 8 | 1282 | 66 | ||
NM_166677.3 | NP_726461.1 | 8 - 9 | 1564 | 66 | ||
GenBank mRNA | AY862157.1 | - | 8 - 9 | 1534 | 66 | |
BT003795.1 | - | 7 - 8 | 2019 | 66 | ||
BT011382.1 | - | 8 - 9 | 1532 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |