Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202070.1 | NP_001188999.1 | 3 - 4 | 1016 | 88 | |
NM_001202071.1 | NP_001189000.1 | 3 - 4 | 1016 | 88 | ||
NM_001202072.2 | NP_001189001.1 | 3 - 4 | 921 | 88 | ||
NM_001202073.1 | NP_001189002.1 | 3 - 4 | 1016 | 88 | ||
NM_001202074.2 | NP_001189003.1 | 3 - 4 | 1007 | 88 | ||
NM_001202075.1 | NP_001189004.1 | 3 - 4 | 1016 | 88 | ||
NM_001202076.1 | NP_001189005.1 | 3 - 4 | 1016 | 88 | ||
NM_001259570.2 | NP_001246499.1 | 3 - 4 | 1007 | 88 | ||
NM_079128.4 | NP_523852.3 | 3 - 4 | 1007 | 88 | ||
NM_166683.2 | NP_726473.2 | 3 - 4 | 1007 | 88 | ||
GenBank mRNA | AY118405.1 | - | 3 - 4 | 1003 | 88 | |
BT021303.1 | - | 3 - 4 | 1003 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |