Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259573.1 | NP_001246502.1 | 2 - 3 | 596 | 69 | |
NM_138121.2 | NP_611965.2 | 2 - 3 | 596 | 69 | ||
NM_166686.1 | NP_726481.1 | 2 - 3 | 596 | 69 | ||
NM_166687.1 | NP_726482.1 | 2 - 3 | 596 | 69 | ||
GenBank mRNA | BT125713.1 | - | 2 - 3 | 562 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |