Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169829.3 | NP_001163300.1 | 6 - 7 | 1624 | 107 | |
NM_166705.3 | NP_726508.1 | 6 - 7 | 1624 | 107 | ||
GenBank mRNA | AY069169.1 | - | 6 - 7 | 1597 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |