Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014687.4 | NP_001014687.3 | 13 - 14 | 2951 | 85 | |
NM_001014688.4 | NP_001014688.3 | 13 - 14 | 2951 | 85 | ||
NM_001014689.5 | NP_001014689.3 | 13 - 14 | 2951 | 85 | ||
NM_001038714.4 | NP_001033803.3 | 13 - 14 | 2951 | 85 | ||
NM_001201585.2 | NP_001188514.1 | 13 - 14 | 2917 | 85 | ||
NM_001201586.2 | NP_001188515.1 | 12 - 13 | 2833 | 85 | ||
NM_001201587.2 | NP_001188516.1 | 13 - 14 | 2951 | 85 | ||
NM_001201588.2 | NP_001188517.1 | 13 - 14 | 2869 | 85 | ||
NM_001201589.2 | NP_001188518.1 | 13 - 14 | 2951 | 85 | ||
NM_166754.6 | NP_726564.5 | 13 - 14 | 2951 | 85 | ||
NM_166755.6 | NP_726565.5 | 13 - 14 | 2869 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |