Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephrin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143670.5 | NP_651927.1 | 3 - 4 | 1236 | 94 | |
NM_166770.2 | NP_726583.1 | 3 - 4 | 1339 | 94 | ||
NM_166771.2 | NP_726584.1 | 3 - 4 | 1544 | 94 | ||
GenBank mRNA | AF216287.1 | - | 3 - 4 | 1184 | 94 | |
BT005199.1 | - | 3 - 4 | 1300 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |