Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CaMKII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014696.2 | NP_001014696.1 | 13 - 14 | 1580 | 122 | |
NM_001169360.2 | NP_001162831.1 | 12 - 13 | 1981 | 122 | ||
NM_001169361.2 | NP_001162832.1 | 15 - 16 | 1518 | 122 | ||
NM_001169362.2 | NP_001162833.1 | 14 - 15 | 1455 | 122 | ||
NM_079896.4 | NP_524635.3 | 12 - 13 | 1502 | 122 | ||
NM_166810.5 | NP_726633.2 | 12 - 13 | 1332 | 122 | ||
NM_166811.2 | NP_726634.1 | 13 - 14 | 1473 | 122 | ||
NM_166812.3 | NP_726635.2 | 14 - 15 | 1622 | 122 | ||
NM_166813.3 | NP_726636.2 | 13 - 14 | 1559 | 122 | ||
GenBank mRNA | BT050453.1 | - | 12 - 13 | 1920 | 122 | |
M74583.1 | - | 12 - 13 | 1456 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |