Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258501.3 | NP_001245430.1 | 4 - 5 | 989 | 83 | |
NM_143758.4 | NP_652015.1 | 4 - 5 | 1092 | 83 | ||
NM_166815.3 | NP_726638.1 | 3 - 4 | 819 | 83 | ||
NM_166816.3 | NP_726639.1 | 4 - 5 | 1010 | 83 | ||
NM_166819.3 | NP_726642.1 | 5 - 6 | 1211 | 83 | ||
GenBank mRNA | AF219948.1 | - | 4 - 5 | 1045 | 83 | |
AY069355.1 | - | 4 - 5 | 982 | 83 | ||
BT150084.1 | - | 3 - 4 | 788 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |