Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cals | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272157.1 | NP_001259086.1 | 9 - 10 | 2805 | 124 | |
NM_001297793.1 | NP_001284722.1 | 9 - 10 | 2808 | 124 | ||
NM_079893.3 | NP_524632.1 | 9 - 10 | 2805 | 124 | ||
NM_166823.2 | NP_726646.1 | 9 - 10 | 2808 | 124 | ||
GenBank mRNA | AJ289018.1 | - | 9 - 10 | 2771 | 124 | |
AY121644.1 | - | 9 - 10 | 2773 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |