Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057682.4 | NP_477030.1 | 6 - 7 | 1787 | 60 | |
NM_166835.4 | NP_726659.1 | 6 - 7 | 1787 | 60 | ||
GenBank mRNA | AY069078.1 | - | 7 - 8 | 2155 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |