Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ewg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014707.2 | NP_001014707.1 | 2 - 3 | 1507 | 63 | |
NM_001014708.2 | NP_001014708.1 | 2 - 3 | 1507 | 63 | ||
NM_001014709.2 | NP_001014709.1 | 2 - 3 | 1507 | 63 | ||
NM_001014710.2 | NP_001014710.1 | 2 - 3 | 1507 | 63 | ||
NM_001014711.2 | NP_001014711.1 | 2 - 3 | 1507 | 63 | ||
NM_001014712.2 | NP_001014712.1 | 2 - 3 | 1507 | 63 | ||
NM_001272163.1 | NP_001259092.1 | 2 - 3 | 1507 | 63 | ||
NM_001272164.1 | NP_001259093.1 | 2 - 3 | 1507 | 63 | ||
NM_057544.4 | NP_476892.4 | 2 - 3 | 1507 | 63 | ||
NM_166836.2 | NP_726660.3 | 1 - 2 | 2906 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |