Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ewg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014708.2 | NP_001014708.1 | 6 - 7 | 2101 | 81 | |
NM_001014710.2 | NP_001014710.1 | 8 - 9 | 2637 | 81 | ||
NM_001014711.2 | NP_001014711.1 | 7 - 8 | 2563 | 81 | ||
NM_001014712.2 | NP_001014712.1 | 7 - 8 | 2175 | 81 | ||
NM_166836.2 | NP_726660.3 | 6 - 7 | 3962 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |