Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201593.2 | NP_001188522.1 | 9 - 10 | 3842 | 54 | |
NM_001272197.1 | NP_001259126.1 | 10 - 11 | 3782 | 54 | ||
NM_001297842.1 | NP_001284771.1 | 10 - 11 | 3695 | 54 | ||
NM_130544.4 | NP_569900.3 | 8 - 9 | 2642 | 54 | ||
NM_166875.3 | NP_726723.2 | 9 - 10 | 3525 | 54 | ||
NM_166876.3 | NP_726724.1 | 11 - 12 | 3737 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |