Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmdar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014714.2 | NP_001014714.1 | 5 - 6 | 2603 | 63 | |
NM_001014715.2 | NP_001014715.1 | 4 - 5 | 1897 | 63 | ||
NM_001014716.2 | NP_001014716.1 | 4 - 5 | 1554 | 63 | ||
NM_001169164.1 | NP_001162635.1 | 4 - 5 | 1611 | 63 | ||
NM_001169165.1 | NP_001162636.1 | 4 - 5 | 1621 | 63 | ||
NM_001169166.1 | NP_001162637.1 | 5 - 6 | 2604 | 63 | ||
NM_001297859.1 | NP_001284788.1 | 5 - 6 | 2604 | 63 | ||
GenBank mRNA | AY050490.1 | - | 4 - 5 | 1554 | 63 | |
AY050491.1 | - | 4 - 5 | 1897 | 63 | ||
AY616144.1 | - | 4 - 5 | 1878 | 63 | ||
AY616145.1 | - | 3 - 4 | 2136 | 63 | ||
AY616146.1 | - | 4 - 5 | 1621 | 63 | ||
AY616147.1 | - | 4 - 5 | 1611 | 63 | ||
AY616148.1 | - | 5 - 6 | 2604 | 63 | ||
AY616149.1 | - | 5 - 6 | 2604 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |