Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmdar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014714.2 | NP_001014714.1 | 9 - 10 | 3705 | 91 | |
NM_001014715.2 | NP_001014715.1 | 8 - 9 | 2999 | 91 | ||
NM_001014716.2 | NP_001014716.1 | 8 - 9 | 2656 | 91 | ||
NM_001169164.1 | NP_001162635.1 | 8 - 9 | 2713 | 91 | ||
NM_001169165.1 | NP_001162636.1 | 8 - 9 | 2723 | 91 | ||
NM_001169166.1 | NP_001162637.1 | 9 - 10 | 3706 | 91 | ||
NM_001297859.1 | NP_001284788.1 | 9 - 10 | 3706 | 91 | ||
GenBank mRNA | AY050490.1 | - | 8 - 9 | 2656 | 91 | |
AY050491.1 | - | 8 - 9 | 2999 | 91 | ||
AY616144.1 | - | 8 - 9 | 2980 | 91 | ||
AY616145.1 | - | 7 - 8 | 3238 | 91 | ||
AY616146.1 | - | 8 - 9 | 2723 | 91 | ||
AY616147.1 | - | 8 - 9 | 2713 | 91 | ||
AY616148.1 | - | 9 - 10 | 3706 | 91 | ||
AY616149.1 | - | 9 - 10 | 3706 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |