Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201603.2 | NP_001188532.1 | 9 - 10 | 2724 | 67 | |
NM_001201604.2 | NP_001188533.1 | 9 - 10 | 2934 | 67 | ||
NM_001272235.1 | NP_001259164.1 | 10 - 11 | 2935 | 67 | ||
NM_001272236.1 | NP_001259165.1 | 9 - 10 | 2787 | 67 | ||
NM_001272237.1 | NP_001259166.1 | 9 - 10 | 2769 | 67 | ||
NM_001272238.1 | NP_001259167.1 | 9 - 10 | 2784 | 67 | ||
NM_058136.4 | NP_477484.2 | 9 - 10 | 2929 | 67 | ||
NM_058137.6 | NP_477485.1 | 9 - 10 | 2992 | 67 | ||
NM_166920.2 | NP_726784.1 | 9 - 10 | 2929 | 67 | ||
GenBank mRNA | AY069615.1 | - | 9 - 10 | 2842 | 67 | |
BT126008.1 | - | 10 - 11 | 2936 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |