Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169695.2 | NP_001163166.1 | 6 - 7 | 1659 | 65 | |
NM_001299547.1 | NP_001286476.1 | 6 - 7 | 1647 | 65 | ||
NM_079034.5 | NP_523758.3 | 6 - 7 | 1706 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |