Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274097.2 | NP_001261026.1 | 4 - 5 | 801 | 83 | |
NM_079039.4 | NP_523763.2 | 4 - 5 | 801 | 83 | ||
GenBank mRNA | AF439752.1 | - | 3 - 4 | 534 | 83 | |
AY051579.1 | - | 4 - 5 | 821 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |