Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or56a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079072.2 | NP_523796.2 | 2 - 3 | 726 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |