Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qkr58E-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299793.1 | NP_001286722.1 | 2 - 3 | 881 | 121 | |
NM_001299794.1 | NP_001286723.1 | 2 - 3 | 881 | 121 | ||
NM_079085.5 | NP_523809.2 | 2 - 3 | 881 | 121 | ||
GenBank mRNA | AY118745.1 | - | 2 - 3 | 905 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |