Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qkr58E-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299795.1 | NP_001286724.1 | 2 - 3 | 1413 | 76 | |
NM_001299796.1 | NP_001286725.1 | 2 - 3 | 1164 | 76 | ||
NM_079086.3 | NP_523810.2 | 2 - 3 | 1164 | 76 | ||
GenBank mRNA | BT021292.1 | - | 2 - 3 | 1160 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |