Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nvy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274235.1 | NP_001261164.1 | 5 - 6 | 1946 | 61 | |
NM_001274236.1 | NP_001261165.1 | 5 - 6 | 1953 | 61 | ||
NM_001274237.2 | NP_001261166.1 | 5 - 6 | 2275 | 61 | ||
NM_079117.4 | NP_523841.3 | 5 - 6 | 2275 | 61 | ||
GenBank mRNA | BT004898.1 | - | 5 - 6 | 2251 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |