Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274238.1 | NP_001261167.1 | 1 - 2 | 363 | 62 | |
NM_001274239.1 | NP_001261168.1 | 2 - 3 | 380 | 62 | ||
NM_079121.3 | NP_523845.2 | 1 - 2 | 363 | 62 | ||
GenBank mRNA | AY089283.1 | - | 1 - 2 | 397 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |